Follicle dermal papilla cell culture medium
Supplier: PROMOCELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C-26502 C-26501 C-26500 C-26505
C-26502EA 241 GBP
C-26502 C-26501 C-26500 C-26505
Follicle dermal papilla cell culture medium
Cell Culture Media
PromoCell Follicle Dermal Papilla Cell Culture Medium is a low-serum medium. The combination of low serum and growth factors leads to excellent proliferation of human follicle dermal papilla cells in vitro.
Delivery information: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. Medium Kit consists of 1× 500 ml bottle basal medium and 1× SupplementPack (individual vials with pre-measured supplements).