Anti-GPR119 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NLS548 NLS548SS
NOVUNLS548EA 509 GBP
NOVUNLS548 NOVUNLS548SS
Anti-GPR119 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal GPR119 Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100.

Type: Primary
Antigen: GPR119 (G protein-coupled receptor 119)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol GPR119 GPR119
Environmentally Preferable
Antigen name G Protein-Coupled Receptor 119 G Protein-Coupled Receptor 119
Antigen synonyms G-protein coupled receptor 2|RP1-20I3.4|GPCR2N: MGC119957N: hGPCR2 G-protein coupled receptor 2|RP1-20I3.4|GPCR2N: MGC119957N: hGPCR2
Antibody type Primary Primary
Clonality Polyclonal Polyclonal
Conjugation Unconjugated Unconjugated
Reactivity Human Human
Host Rabbit Rabbit
Gene ID 139760 139760
Isotype IgG IgG
ImmunoChemistry Yes Yes
ImmunoFluorescence Yes Yes
Immunogen A synthetic peptide made to an internal region of the human GPR119 protein (within residues 150-250). [Swiss-Prot Q8TDV5] A synthetic peptide made to an internal region of the human GPR119 protein (within residues 150-250). [Swiss-Prot Q8TDV5]
Purification Immunogen affinity purified Immunogen affinity purified
Concentration 1 mg/ml 1 mg/ml
Storage buffer PBS and 30% Glycerol PBS and 30% Glycerol
Storage temperature Cool Cool
Shipping temperature Ice
Tested applications Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR