Anti-SHH Mouse Monoclonal Antibody [clone: 5H4]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NBP2-22126 NBP2-22126SS
NOVUNBP2-22126EA 543 GBP
NOVUNBP2-22126 NOVUNBP2-22126SS
Anti-SHH Mouse Monoclonal Antibody [clone: 5H4]
Antibodies
Type: Primary
Antigen: SHH (sonic hedgehog)
Clonality: monoclonal
Clone: 5H4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol SHH
Environmentally Preferable
Antigen name Sonic Hedgehog
Antigen synonyms TPT|HLP3|HHG1|SMMCI|HPE3|MCOPCB5|TPTPS
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 5H4
Reactivity
Host Mouse
Gene ID 6469
Isotype IgG1
Western blot Yes
ELISA Yes
Flow cytometry Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR