Anti-HPx1 Mouse Monoclonal Antibody [clone: HIC0-3B3]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NBP1-18951 NBP1-18951SS
NOVUNBP1-18951EA 539 GBP
NOVUNBP1-18951 NOVUNBP1-18951SS
Anti-HPx1 Mouse Monoclonal Antibody [clone: HIC0-3B3]
Antibodies
Type: Primary
Antigen: HPx1
Clonality: monoclonal
Clone: HIC0-3B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol HPx1
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone HIC0-3B3
Reactivity Human
Host Mouse
Isotype IgG1
ImmunoFluorescence Yes
Flow cytometry Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR