Anti-BECN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB500-266 NB500-266SS
NOVUNB500-266EA 539 GBP
NOVUNB500-266 NOVUNB500-266SS
Anti-BECN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol BECN1
Antigen name Beclin 1
Environmentally Preferable
Antigen synonyms Coiled-coil myosin-like BCL2-interacting protein|APG 6|VPS30|Beclin1 autophagy|ATG6|CTIP-1 COUP-TF interacting protein 1|Protein GT197
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 8678
Isotype
Western blot Yes
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR