Anti-CENPF Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB500-101 NB500-101SS
NOVUNB500-101EA 670 GBP
NOVUNB500-101 NOVUNB500-101SS
Anti-CENPF Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CENPF (centromere protein F, 350/400kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR