Anti-COX IV Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB110-39115 NB110-39115SS
NOVUNB110-39115EA 643 GBP
NOVUNB110-39115 NOVUNB110-39115SS
Anti-COX IV Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COX IV
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR