Anti-TRA Mouse Monoclonal Antibody [clone: TRA-1-60(R)]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB100-730 NB100-730SS
NOVUNB100-730EA 668 GBP
NOVUNB100-730 NOVUNB100-730SS
Anti-TRA Mouse Monoclonal Antibody [clone: TRA-1-60(R)]
Antibodies
Type: Primary
Antigen: TRA (T cell receptor alpha locus)
Clonality: monoclonal
Clone: TRA-1-60(R)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol TRA-1-60-R
Antigen name T cell receptor alpha locus
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone TRA-1-60(R)
Reactivity Human
Host Mouse
Isotype IgM
Western blot Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Amino acid number 1 to 60
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR