Anti-IgG Goat Polyclonal Antibody (CF™ 543)

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20317 20317-1
20317.EA 286 GBP
20317. 20317-1
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Guinea pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR