Anti-IgG1 Goat Polyclonal Antibody (CF™ 350)

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20245 20245-1
20245.EA 367 GBP
20245. 20245-1
Anti-IgG1 Goat Polyclonal Antibody (CF™ 350)
Antibodies
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: CF® 350 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR