Anti-IgG Chicken Polyclonal Antibody (CF™ 633)

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20227 20227-1
20227.EA 286 GBP
20227. 20227-1
Anti-IgG Chicken Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Chicken
Isotype: none
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR