Anti-Biotin Mouse Monoclonal Antibody (CF®CF405S)

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20203 20203-1
20203.EA 342 GBP
20203. 20203-1
Anti-Biotin Mouse Monoclonal Antibody (CF®CF405S)
Antibodies
Type: Tag
Antigen: Biotin (biotin)
Clonality: monoclonal
Clone:
Conjugation: CF® CF405S Dye
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR