Anti-Retinoic Acid Receptor beta Rabbit Monoclonal Antibody [clone: EPR2017]

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB124701-100 AB124701-1ML
ABCAAB124701-100EA 483 GBP
ABCAAB124701-100 ABCAAB124701-1ML
Anti-Retinoic Acid Receptor beta Rabbit Monoclonal Antibody [clone: EPR2017]
Antibodies
Rabbit monoclonal [EPR2017] to Retinoic Acid Receptor beta.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Retinoic Acid Receptor beta
Clonality: Monoclonal
Clone: EPR2017
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR