Anti-Vitamin D Receptor Rabbit Monoclonal Antibody [clone: EPR4552]

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB109234-100 AB109234-1ML
ABCAAB109234-100EA 414 GBP
ABCAAB109234-100 ABCAAB109234-1ML
Anti-Vitamin D Receptor Rabbit Monoclonal Antibody [clone: EPR4552]
Antibodies
Rabbit monoclonal [EPR4552] to Vitamin D Receptor - ChIP Grade.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Vitamin D Receptor
Clonality: Monoclonal
Clone: EPR4552
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR