Anti-HLA-A Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB135651-100 ORB135651-200 ORB135651-50
BIRBORB135651-100EA 0 GBP
BIRBORB135651-100 BIRBORB135651-200 BIRBORB135651-50
Anti-HLA-A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HLA-A (major histocompatibility complex, class I, A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR