Anti-CCNB1 Rabbit polyclonal antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB135187-100 ORB135187-200 ORB135187-50
BIRBORB135187-100EA 0 GBP
BIRBORB135187-100 BIRBORB135187-200 BIRBORB135187-50
Anti-CCNB1 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: CCNB1 (cyclin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR