Anti-SDC4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB128949-100 ORB128949-200
BIRBORB128949-100EA 0 GBP
BIRBORB128949-100 BIRBORB128949-200
Anti-SDC4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SDC4 (syndecan 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR