Anti-SERC3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB128712-100 ORB128712-200
BIRBORB128712-100EA 0 GBP
BIRBORB128712-100 BIRBORB128712-200
Anti-SERC3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SERC3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR