Anti-TRIM38 Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB127301-100 ORB127301-200
BIRBORB127301-100EA 0 GBP
BIRBORB127301-100 BIRBORB127301-200
Anti-TRIM38 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRIM38 (tripartite motif containing 38)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR