Anti-QORX Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB126451-100 ORB126451-200
BIRBORB126451-100EA 0 GBP
BIRBORB126451-100 BIRBORB126451-200
Anti-QORX Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: QORX
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR