Anti-GNL3L Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB125785-100 ORB125785-200
BIRBORB125785-100EA 0 GBP
BIRBORB125785-100 BIRBORB125785-200
Anti-GNL3L Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GNL3L (guanine nucleotide binding protein-like 3 (nucleolar)-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR