Anti-CSF2RA Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB125623-100 ORB125623-200
BIRBORB125623-100EA 0 GBP
BIRBORB125623-100 BIRBORB125623-200
Anti-CSF2RA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CSF2RA (colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR