Anti-RHBDD3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB101172-100 ORB101172-200
BIRBORB101172-100EA 0 GBP
BIRBORB101172-100 BIRBORB101172-200
Anti-RHBDD3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200

Type: Primary
Antigen: RHBDD3 (rhomboid domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR