Anti-WAS Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB48949-100 ORB48949-200
BIRBORB48949-100EA 0 GBP
BIRBORB48949-100 BIRBORB48949-200
Anti-WAS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: WAS (Wiskott-Aldrich syndrome)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR