Anti-SELP Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB12111-100 ORB12111-50
BIRBORB12111-100EA 0 GBP
BIRBORB12111-100 BIRBORB12111-50
Anti-SELP Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
FACS: 1:50-100

Type: Primary
Antigen: SELP (selectin P (granule membrane protein 140kDa, antigen CD62))
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR