Anti-PAFAH1B2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB1109-100 ORB1109-200
BIRBORB1109-100EA 0 GBP
BIRBORB1109-100 BIRBORB1109-200
Anti-PAFAH1B2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: PAFAH1B2 (platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR