Anti-PVRL1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB1021-100 ORB1021-200
BIRBORB1021-100EA 0 GBP
BIRBORB1021-100 BIRBORB1021-200
Anti-PVRL1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: PVRL1 (poliovirus receptor-related 1 (herpesvirus entry mediator C))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR