Waste container, SHARPSGUARD® eco pharmi
Supplier: DANIELS HEALTHCARE LIMITED
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DD210 DD205
DANIDD210EA 190 GBP
DANIDD210 DANIDD205
Waste container, SHARPSGUARD® eco pharmi
Bins Waste Bins
SHARPSGUARD® eco pharmi are ideal general-purpose waste containers for the disposal of pharmaceutical waste in original packaging, such as unused pills, no sharps.
  • Aperture in the lid which can be closed temporarily to prevent accidental spillage
  • Sustainable: Base, lid and strap are made from 100% recycled plastic
  • With handle
These containers are for disposal by incineration or other authorised process and are designed for the consignment of waste categorised by EWC codes: 18-01-09 & 18-02-08.
Order Now
SPECIFICATIONS
Environmentally Preferable