Cleanroom gloves, nitrile, sterile, Nitrex 600
Supplier: DISTRIBUTOR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GN38-7.5 GN38-8.5
DIUKGN38-7.5EA 1210 GBP
DIUKGN38-7.5 DIUKGN38-8.5
Cleanroom gloves, nitrile, sterile, Nitrex 600
Gloves
These nitrile butadiene rubber gloves are ideal for use in clinics, laboratories and cleanroom environments.
  • Sterile, powder-free, smooth finish
  • Chlorination on donning surface
  • Latex-free
  • Sterilised by gamma irradiation (min. 25 kGy)
EN 455-1/-2/-3; AQL 1,5
Certifications: EN 455-1/-2/-3; AQL 1,5
Packaging: 100 pairs per case.