Cryostats, HM525 NX
Supplier: Epredia
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
956650
720-2710EA 0 GBP
720-2710
Cryostats, HM525 NX
Microtomes and Microtome Assemblies Microtomes
The HM525 NX is a high performance routine cryostat with intuitive software and touch screen. The front of the microtome stops build up of debris behind the microtome making it easier to clean.
  • Operator safety and minimal service downtime
  • Cooled specimen positions (27) including four fast-freezing stations to −57 °C to limit ice crystal formation
  • Additional knee space supports comfortable seated operation
  • Ten minute specimen fast-freeze function can be accessed from the menu
Optional UV disinfection: High intensity UV light delivers disinfection for peace of mind and enhanced germicidal activity; immediate or scheduled operation; alert screen indicates incomplete or interrupted disinfection cycle and adjustable disinfection time setting up to 300 min.