Analytical balances, GR series
Supplier: AnD Instruments Ltd (A&D)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GR-200 GR-202
ANDAGR-200EA 0 GBP
ANDAGR-200 611-3251
Analytical balances, GR series
Balances and Scales
A series of analytical balances from A&D for advanced, accurate and reliable weighing. Applications include counting with Automatic Counting Accuracy Improvement (ACAI), percentage weighing, below balance weighing and density determination.
  • One touch digital calibration using internal motor driven masses with automatic self calibration on ambient temperature changes
  • 11 weighing units plus counting and percentage
  • Two layer housing construction minimises the negative effects of ambient temperature changes
  • Sealed keypad protecting against spills and dust
  • Built-in underhook capability
  • RS232 bi-directional interface
The data memory function allows 200 sets of weighing data to be stored (or 100 sets of weighing data with time and date, or 50 sets of calibration data). GLP/GMP compliant data output, including calibration weight, time and date.
Ordering information: Please contact your local Avantor sales office for details on other accessories including LAN-Ethernet interface with WinCT-Plus software, density determination kits, analytical pans, remote display and calibration weights.
Delivery information: Supplied with AC adapter, instruction manual.