Refrigerated chromatography cabinets, Eco Pro G2
Supplier: FOSTER REFRIGERATION
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EP700G EP1440G-SS
471-1107EA 0 GBP
471-1107 471-1104
Refrigerated chromatography cabinets, Eco Pro G2
Refrigerators
The EcoPro G2 has been loaded with new design, quality, reliability and performance features.
  • Thick and robust cabinet design with advanced thermal efficiency
  • 'Smartphone style' touch sensitive hidden-until-lit control panel with unique high visibility temperature display combines sophistication with functionality
  • Ergonomic door and handle design for ease of opening and cleaning
  • With 'Fluid Food-Fresh Dynamics' modelling: advanced Circular II air distribution system maximises efficient operation even when shelves are fully loaded