Air inlet tubes, straight
Supplier: SMITH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8SMCS0 8SMCS1 8SMCS2 8SMCS3
SMIT8SMCS0EA 105 GBP
SMIT8SMCS0 SMIT8SMCS1 SMIT8SMCS2 SMIT8SMCS3
Air inlet tubes, straight
Tubes Gas Inlet Tubes
Borosilicate glass. Straight, with cone.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes