Powder funnels
Supplier: SMITH SCIENTIFIC LTD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8SPF/B14 8SPF/B19 8SPF/B24 8SPF/B29 8SPF/B34
SMIT8SPF/B14EA 118 GBP
SMIT8SPF/B14 SMIT8SPF/B19 SMIT8SPF/B24 SMIT8SPF/B29 SMIT8SPF/B34
Powder funnels
Funnels Filtration Funnels Funnels with Conical Joints
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Stem with cone.