Cannon-Fenske viscometers
Supplier: STANHOPE SETA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11634/01 11634/02 11634/03 11634/04 11634/05 11634/06 11634/07 11634/08 11634/09 11634/11 11634/12
STAN11634/01EA 609 GBP
STAN11634/01 STAN11634/02 STAN11634/03 STAN11634/04 STAN11634/05 STAN11634/06 STAN11634/07 STAN11634/08 STAN11634/09 STAN11634/11 STAN11634/12
Cannon-Fenske viscometers
Viscometers Capillary Viscometers
Capillary viscometers for transparent liquids.
Calibration with constant at 40 °C.
Certifications: ASTM D445 and D446, ISO 3104 and 3105.