Dishes, evaporating
Supplier: Smith Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SMIT179-95/170EA 0 GBP
SMIT179-95/170 SMIT179-80/90 SMIT179/140 SMIT179/95 SMIT179/80 SMIT179/60
Dishes, evaporating
Trays/Dishes/Pots Evaporating Basins/Dishes
Grade 3.3 borosilicate glass, with spout.
  • Excellent chemical resistance
  • High temperature resistance up to +500 °C
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide range of applications. With flat bottom and matt labelling area.