Dishes, crystallising
Supplier: SMITH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
174/115 174/140 174/190 174/230 174/40 174/50 174/60 174/70 174/80 174/95 175/115 175/140 175/190 175/230 175/50 175/60 175/70 175/80 175/95 174-230/3500
SMIT174/115EA 84.2 GBP
SMIT174/115 SMIT174/140 SMIT174/190 SMIT174/230 SMIT174/40 SMIT174/50 SMIT174/60 SMIT174/70 SMIT174/80 SMIT174/95 SMIT175/115 SMIT175/140 SMIT175/190 SMIT175/230 SMIT175/50 SMIT175/60 SMIT175/70 SMIT175/80 SMIT175/95 SMIT174-230/3500
Dishes, crystallising
Trays/Dishes/Pots Crystallising Dishes
Crystallising dishes with or without spout, made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes