4-Bromotetrahydropyran

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromooxane, 4-Bromotetrahydro-2H-pyran

AS20466G1 AS20466G5 AS20466G25
ACTIAS20466G1EA 8 GBP
ACTIAS20466G1 ACTIAS20466G5 ACTIAS20466G25
4-Bromotetrahydropyran
4-Bromotetrahydropyran
Formula: C₅H₉BrO
MW: 165.0300 g/mol
MDL Number: MFCD07371501
CAS Number: 25637-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR