4-(Bromomethyl)tetrahydropyran

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS1354G1 AS1354G25 AS1354G5
ACTIAS1354G1EA 0 GBP
ACTIAS1354G1 ACTIAS1354G25 ACTIAS1354G5
4-(Bromomethyl)tetrahydropyran
4-(Bromomethyl)tetrahydropyran
Formula: C₆H₁₁BrO
MW: 179.0549 g/mol
MDL Number: MFCD03788561
CAS Number: 125552-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR