4,4'-Dimethyl-2,2'-bipyridyl
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4,4'-Dimethyl-2,2'-bipyridine
AS23507G25
AS23507G5
ACTIAS23507G25EA
0
GBP
ACTIAS23507G25
ACTIAS23507G5
4,4'-Dimethyl-2,2'-bipyridyl
4,4'-Dimethyl-2,2'-bipyridyl
Formula:
C₁₂H₁₂N₂ MW: 184.24 g/mol |
MDL Number:
MFCD00006441 CAS Number: 1134-35-6 EINECS: 214-483-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...