3-Bromo-4-fluorophenol 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H61702.03
H61702.06
H61702.03EA
18.3
GBP
H61702.03
H61702.06
3-Bromo-4-fluorophenol 97%
3-Bromo-4-fluorophenol
Purity: 98%
CAS No.: 27407-11-0
Molecular Formula: C6H4BrFO
Formula Weight: 191.00
Physical Form: Crystalline
Appearance: Yellow-green
UN No.: 2811
MDL No.: MFCD03425884
Formula:
C₆H₄BrFO Storage Temperature: Ambient |
MDL Number:
MFCD03425884 CAS Number: 27407-11-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...