Toluene
Supplier: LAB 3
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
RG2025PE
RG2025
LBNORG2025PEEA
0
GBP
LBNORG2025PE
LBNORG2025
Toluene
Toluene
Formula:
C₆H₅CH₃ Boiling Pt: 110.6 °C (1013 hPa) Melting Pt: –95 °C Density: 0.867 g/cm³ (20 °C) Flash Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3,II REACH: 05-2114615130-69 Merck Index: 13,09607 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...