1-Benzyloxy-4-fluorobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Benzyloxy)-4-fluorobenzene

OT-0795-5G OT-0795-25G OT-0795-100G
COBBOT-0795-5GEA 0 GBP
COBBOT-0795-5G COBBOT-0795-25G COBBOT-0795-100G
1-Benzyloxy-4-fluorobenzene
1-Benzyloxy-4-fluorobenzene
Formula: C₁₃H₁₁FO
MW: 202.23
MDL Number: MFCD03788273
CAS Number: 370-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR