2,4,5-Trichloroanisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0706-5G OT-0706-1G
COBBOT-0706-5GEA 0 GBP
COBBOT-0706-5G COBBOT-0706-1G
2,4,5-Trichloroanisole
2,4,5-Trichloroanisole
Formula: C₇H₅Cl₃O
MDL Number: MFCD00129716
CAS Number: 6130-75-2
EINECS: 228-099-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR