2-Methoxy-5-(trifluoromethyl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7349-1G PY-7349-25G PY-7349-5G
COBBPY-7349-1GEA 0 GBP
COBBPY-7349-1G COBBPY-7349-25G COBBPY-7349-5G
2-Methoxy-5-(trifluoromethyl)pyridine
2-Methoxy-5-(trifluoromethyl)pyridine
Formula: C₇H₆F₃NO
MDL Number: MFCD00153203
CAS Number: 175277-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR