3-Cyclopentene-1-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: cyclopent-3-ene-1-carboxylic acid, Cyclopent-3-enecarboxylic acid

AS21027G100 AS21027G25
ACTIAS21027G100EA 0 GBP
ACTIAS21027G100 ACTIAS21027G25
3-Cyclopentene-1-carboxylic acid
3-Cyclopentene-1-carboxylic acid
Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00101764
CAS Number: 7686-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR