4-Hydroxy-1-(2-nitro-5-(trifluoromethyl)phenyl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2096-5G HC-2096-1G
COBBHC-2096-5GEA 0 GBP
COBBHC-2096-5G COBBHC-2096-1G
4-Hydroxy-1-(2-nitro-5-(trifluoromethyl)phenyl)piperidine
4-Hydroxy-1-(2-nitro-5-(trifluoromethyl)phenyl)piperidine
Formula: C₁₂H₁₃F₃N₂O₃
MDL Number: MFCD11504892
CAS Number: 1072944-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR