(S)-(+)-Ethyl-3-piperidinecarboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl L-nipecotate, Ethyl (S)-(+)-3-Piperidinecarboxylate, (S)-Ethyl piperidine-3-carboxylate

AS24053G100 AS24053G25 AS24053G5
ACTIAS24053G100EA 265 GBP
ACTIAS24053G100 ACTIAS24053G25 ACTIAS24053G5
(S)-(+)-Ethyl-3-piperidinecarboxylate
(S)-(+)-Ethyl 3-piperidinecarboxylate
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
MDL Number: MFCD00792499
CAS Number: 37675-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR