2-Bromo-1-iodo-4-(trifluoromethoxy)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-(trifluoromethoxy)iodobenzene

OT-0738-25G OT-0738-5G
COBBOT-0738-25GEA 0 GBP
COBBOT-0738-25G COBBOT-0738-5G
2-Bromo-1-iodo-4-(trifluoromethoxy)benzene
2-Bromo-1-iodo-4-(trifluoromethoxy)benzene
Formula: C₇H₃BrF₃IO
MDL Number: MFCD07369009
CAS Number: 883546-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR