1-Bromo-2-iodo-4-(trifluoromethoxy)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2988-5G HC-2988-1G
COBBHC-2988-5GEA 0 GBP
COBBHC-2988-5G COBBHC-2988-1G
1-Bromo-2-iodo-4-(trifluoromethoxy)benzene
1-Bromo-2-iodo-4-(trifluoromethoxy)benzene
Formula: C₇H₃BrF₃IO
MW: 366.901 g/mol
MDL Number: MFCD12025452
CAS Number: 1121586-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR